Would you like to log in again?
AdjusTable® Workstation, Stainless Steel Solid Top, Chrome Base, Eagle MHC™
Supplier: Eagle MHC
AdjusTable®
AC3048T AC3060T AC3036T AC2436T AC2448T AC2460T
58202-828EA 1604.71 USD InStock
58202-828 58202-838 58202-832 58202-830 58202-836 58202-834
AdjusTable® Workstation, Stainless Steel Solid Top, Chrome Base, Eagle MHC™
Furniture Tables and Workbenches
16 gauge, Type 304 brushed stainless steel top. All work surface tabletops feature hat channel underneath for added strength.
  • 34.5" (876 mm) working height
  • Easy to add undershelves or overshelves
  • Available in solid or perforated, with brushed or electropolished finish
  • Four 33" (838 mm) chrome posts or stainless steel posts numbered in 1" (25 mm) increments for flexible and level positioning of the base
Base combination available with or without wire undershelf, with chrome or stainless steel H-frame(s) or C-frame(s).
Load capacity: 600 lbs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty