Would you like to log in again?
AdjusTable® Workstations, Stainless Steel Solid Top with Overshelves, Stainless Steel Base, Eagle MHC™
Supplier: Eagle MHC
AdjusTable®
WOAC2460T WOAS3048T WOAS3036T WOAC2448T WOAC2436T WOAS2460T WOAC3060T WOAC3048T WOAC3036T WOAS2436T WOAS2448T WOAS3060T
58203-010EA 4577.23 USD InStock
58203-010 58202-992 58203-000 58202-990 58203-012 58203-002 58202-996 58203-004 58202-994 58203-006 58203-008 58202-998
AdjusTable® Workstations, Stainless Steel Solid Top with Overshelves, Stainless Steel Base, Eagle MHC™
Furniture Tables and Workbenches
16 gauge Type 304 brushed stainless steel top.
  • Base features 1 stainless steel wire shelf and 1 stainless steel plated C-frame
  • Worsurface height: 34.5"
Ordering information: Include two 63" stainless steel posts and two 33" stainless steel posts and two 14" wide stainless steel wire overshelves.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty