Would you like to log in again?
Staticmaster® Static Eliminating Brushes, NRD
Supplier: NRD
Staticmaster
3C500 1C200
58578-028EA 287.5 USD InStock
58578-028 58578-040
Staticmaster® Static Eliminating Brushes, NRD
Brushes
NRD’s static elimination brushes are used in a wide variety of analytical, photographic and medical environments where the elimination of dust on sensitive tools and objects are necessary.
  • Removes dust particles
  • Neutralizes static
The brushes are hand-crafted with natural hair and contain an alpha ionizing strip which neutralizes the static while the soft brushes simultaneously remove dust particles. The neutralization of the surface with the brushes will allow the surface to stay free of dust.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty