Would you like to log in again?
Mat and Tabletop Cleaner, ACL Staticide
Supplier: ACL Staticide
6001 6002
58611-198CS 241.27 USD InStock
58611-198 58611-196
Mat and Tabletop Cleaner, ACL Staticide
Surface Treatments Antistatic Treatments
Formulated to clean tabletops, antistatic mats, and workstations
Removes flux residues, oil, grease, and inorganic contaminants. Perfect for assembly areas where sensitive electronic components are handled. Maintains original surface resistivity of antistatic mats. Nonflammable and safe for any surface.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty