Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Market Forge Sterilmatic® STM-EX/ELX, Steam Pressure Sterilizers, Export Only, Crown Steam

Supplier: Crown Steam
STM-EX STM-ELX
58700-011EA 20774.67 USD InStock
58700-011 58700-010
Market Forge Sterilmatic® STM-EX/ELX, Steam Pressure Sterilizers, Export Only, Crown Steam
Sterilizers Autoclaves (Steam Sterilizers)
Sterilizing cycle is fully automatic, time-controlled and model STM-EX has built-in temperature control which provides 250 °F (121 °C). Model STM-ELX features adjustable temperature control that can be set anywhere from 230 to 250°F (110- 121°C) range.

  • Thermometer 180 to 300 °F (82 to 149 °C)
  • 0 to 30 lbs. steam gauge
  • 0 to 60 min timer
  • Thermostatic steam trap
  • Signal light
  • Flat perforated steam baffles
  • Removable pan supports
  • Adjustable 4 to 6” bullet feet
  • Shipping weight, all models : 77 kg (169 lbs.)

All models have operating controls that are located at the top, front of the Sterilizer. Pressure is automatically and quickly exhausted and the power supply is shut off at the end of the cycle. Slow exhaust can be selected for sterilizing liquids. All models are manual fill, have a safety valve, low water cut-off and include automatic temperature control.

Certifications: CSA certified.

Ordering information: 90 day parts and labor warranty, one-year parts warranty

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

SERVICES


Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty