Would you like to log in again?
Steri-Wrap® II, Propper Manufacturing
Supplier: Propper
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Steri-Wrap®
31205400 31202400 31201500 31203620 31203040
58752-975CS 431.04 USD InStock
58752-975 76334-066 89125-870 76334-064 76334-062
Steri-Wrap® II, Propper Manufacturing
Sterilization Wraps
Single use non-woven fabric sterilization wrap offers greater drapeability than standard wraps.
  • Cellulose based wrap for steam and EO sterilization
  • Efficient microbial barrier, high-level of protection against fluids and particles
  • Highly permeable meaning no wet packs
  • Variety of strength of fabric to meet your needs
Steri-Wrap® is cellulose based sterilization wrap available in three grades for keeping packages safe and secure during the sterilization process. Steri-Wrap® II is light weight wrap designed for great drapeability.
Steri-Wrap® is designed to be virtually lint free with tightly anchored fibers. Wrap is reversible and features excellent liquid repellency. Steri-Wrap® can be used in steam or ethylene oxide sterilization.
Certifications: ISO 11607-1:2019
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty