Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chex-AII® Heat Sealable Pouches, Propper Manufacturing

Supplier: Propper

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Chex-All®
02500800 02501200 02501000 02502400 02501400 02501800
58753-256CS 390.37 USD InStock
58753-256 58753-258 58753-260 58753-250 58753-252 58753-254
Chex-AII® Heat Sealable Pouches, Propper Manufacturing
Bags Sterilization Pouches
Heat-Sealable Chex-All® pouches are premium quality sterile packaging products designed to reduce the cost of sterilization processing.

  • Heat-Sealing Sterilization Pouches for use in steam and ethylene oxide
  • Pre-printed indicator inside and outside the pouch
  • Features triple-sealed paper/plastic film to ensure security of contents
  • Easy-to-read chemical indicator changes from yellow to black in steam
  • Easy-to-read chemical indicator changes from red to green in EO
  • Made of 2.5 mm plastic and 73 to 74 g

Chex-All® pouches provide breathability, a sterile barrier, and easy to read built-in internal indicators (both inside and outside the pouch). Heat-Sealer provides the convenience of custom sizing to meet the individual needs of your instruments. Chex-All® pouches are for use in steam and ethylene oxide, eliminated the need for two sets of pouches. Chex-All® pouches are validated for double pouch packaging.

Certifications: AAMI ST60/ ISO 11140-1, Type 1 Process Indicators.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty