Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Chex-AII® Heat Sealable Pouches, Propper Manufacturing
Supplier: Propper
Chex-All®
02500800 02501200 02501000 02502400 02501400 02501800
58753-256CS 390.37 USD InStock
58753-256 58753-258 58753-260 58753-250 58753-252 58753-254
Chex-AII® Heat Sealable Pouches, Propper Manufacturing
Bags Sterilization Pouches
Heat-Sealable Chex-All® pouches are premium quality sterile packaging products designed to reduce the cost of sterilization processing.
  • Heat-Sealing Sterilization Pouches for use in steam and ethylene oxide
  • Pre-printed indicator inside and outside the pouch
  • Features triple-sealed paper/plastic film to ensure security of contents
  • Easy-to-read chemical indicator changes from yellow to black in steam
  • Easy-to-read chemical indicator changes from red to green in EO
  • Made of 2.5 mm plastic and 73 to 74 g
Chex-All® pouches provide breathability, a sterile barrier, and easy to read built-in internal indicators (both inside and outside the pouch). Heat-Sealer provides the convenience of custom sizing to meet the individual needs of your instruments. Chex-All® pouches are for use in steam and ethylene oxide, eliminated the need for two sets of pouches. Chex-All® pouches are validated for double pouch packaging.
Certifications: AAMI ST60/ ISO 11140-1, Type 1 Process Indicators.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty