Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

MagStir Genie™ Digital Programmable Magnetic Stirrer, 230 V

Supplier: Scientific Industries

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Genie®
SI-0301
58815-256EA 1054.53 USD InStock
58815-256
MagStir Genie™ Digital Programmable Magnetic Stirrer, 230 V
Stirrers Magnetic Stirrers
This versatile magnetic stirrer has a speed range of 10 to 1800 rpm with reverse and interval stirring for applications ranging from gentle stirring for cell culture to aggressive mixing for viscous polymers.

  • 230 V model

Easy-to-read LCD display. Unit features three power levels for various sample viscosities. Manual mode has basic stir, stir reverse, and timer functions. The profiles mode allows users to quickly program speed, ramp rate, stir direction, time (including pause), and power level in up to nine distinct steps. Up to four stirring profiles can be saved. Automatic mode recalls and executes these profiles. Timer ranges from 1 minute to 99 hours.

Ordering information: Two-year manufacturer's warranty. The 230 V model is supplied with an electrical cord without a plug.

Caution: 230 V model. Corresponding model with voltage suitable for use in US (58815-254) is also available, please see separate product page.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty