Would you like to log in again?
Corning® Multi-Position Magnetic Stirrers, Corning
Supplier: Corning
Corning®
440824 440826 440825
58941-280CS 2569.07 USD InStock
58941-280 58941-276 58941-278
Corning® Multi-Position Magnetic Stirrers, Corning
Stirrers Magnetic Stirrers, Multi-position
These multi-position stirrers have integral controls that can accommodate up to nine 250 ml vessels.
  • Compact design saves valuable bench space
  • Offers powerful stirring from 350 to 2000 min⁻¹
  • Rugged lightweight glass filled resin housing
  • Maximum vessel diameter is 75 mm for 9 place unit, 95 mm for 5 place model and 130 mm for 4 place stirrer
Stirring platforms are made of glass-filled resin and are available in four-, five-, or nine-position models. Four-position stirrer 58941-276 features a 12V DC remote control, making it easy to control from outside a fume hood or incubator. Magnets and DC motor combine for smooth stirring, up to 2000 rpm. Speed of all positions is set simultaneously with either the integral knob or the remote control.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty