Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Spinbar® Disposable Stir Bars, Octagon
Supplier: VWR
VWR® Spinbar®
37112-6012 37112-6016 37112-6001 37112-6002
58947-140PK 179.81 USD InStock
58947-140 58947-142 58947-144 58947-146
VWR® Spinbar® Disposable Stir Bars, Octagon
Stirrers Magnetic Stirring Bars
The multifaceted stirring bar surface adds turbulence relative to similar size smooth cylindrical stirring bars.
  • Ideal for single use applications such as mixing radioactive reagents
  • Disposable; eliminates potential for cross-contamination
  • Alnico magnet encapsulated in FDA grade Teflon® PTFE is durable and inert
  • Autoclavable
For single-use application where efficiency and cross-contamination are a concern. Alnico V magnet.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty