Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Circulus™ Magnetic Stir Bars, Color-Coded
Supplier: VWR
VWR® Circulus™
37172-6002 37170-6001 37170-6002 37172-6003 37172-6001 37170-6000 37172-6000 37170-6003
58947-849CS 1030.6 USD InStock
58947-849 58947-929 58947-907 58947-882 58947-860 58947-940 58947-984 58947-962
VWR® Circulus™ Magnetic Stir Bars, Color-Coded
Stirrers Magnetic Stirring Bars
These magnetic stir bars perform well with all problem liquids, highly viscous mixtures, metal particles in fluids, and round (convex) bottom vessels.
  • Excellent self-centering characteristics
  • Fast mixing action with high turbulence
  • Greater stability
  • Less drifting at higher speeds
Available in two lengths, 36 and 54 mm (1 7/16 and 2 1/8"), each is designed as a 8 mm (0.315”) diameter bar with solid, color-coded, 20 mm (0.79") diameter, Teflon® resin end discs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty