Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Spinbar® Stir Bars, Octagon, Assortment
Supplier: Avantor
VWR® Spinbar®
37180-6001
58948-003CS 813.85 USD InStock
58948-003
VWR® Spinbar® Stir Bars, Octagon, Assortment
Stirrers Magnetic Stirring Bars
Assortment of 16 PTFE resin-coated, octagon-shaped magnetic stir bars with pivot ring.
  • Autoclavable
  • PTFE resin coating
  • Most commonly used shape
  • Chemically resistant
  • Great temperature resistance
Chemical and temperature resistance up to 260°C (500°F). Stir bars come in a hinged-top, plastic Spinbox™.
Ordering information: Includes two each of 7x2mm (1/4x1/14"), 10x3mm (1/3x1/9"), 12.7x7.9mm (1/2x5/16"), 15.9x7.9mm (5/8x5/16"), 25.4x9.5mm (1x3/8"), 38.1x9.5mm (11/2x3/8"), 50.8x9.5mm (2x3/8"), 63.5x9.5mm (21/2x3/8") sizes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty