Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Spinbar® Stir Bars, Egg-Shaped, Assortment
Supplier: Avantor
VWR® Spinbar®
37187-6001
58948-078CS 1530.12 USD InStock
58948-078
VWR® Spinbar® Stir Bars, Egg-Shaped, Assortment
Stirrers Magnetic Stirring Bars
This Spinbar® stirring bar collection includes a useful assortment of popular sizes of egg-shaped stirring bars.
  • Excellent for round-bottom flasks
  • Provide their own pivot point without a separate ring or abrupt change in contour
  • Offers minimal contact when used in plastic containers
  • Convenient storage box segregates sizes and styles for easy, no-rummage organization
Designed especially for use in round-bottom vessels, they spin unrestricted by the vessel contour. Alnico magnets encapsulated in FDA grade Teflon® PTFE are durable and inert. Selection of thirteen magnetic stirring bars includes one 76.2 x 19 mm (3 x 3/4") and two each of 19 x 9.5 mm (3/4x3/8"), 25.4 x 12.7 mm (1x1/2"), 31.8 x 15.9 mm (11/4x5/8"), 38.1 x 15.9 mm (11/2x5/8"), 41.3 x 19mm (15/8x3/4"), and 50.8 x 19mm (2x3/4"). Autoclavable.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty