Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

VWR® Spinbar® Magnetic Stir Bars, Octagon

Supplier: VWR
VWR® Spinbar®
37110-6016 37110-6010 37110-6019 37110-6017 37110-6007 37110-6013 37110-6014 37110-6001 37110-6015 37110-6022 37110-6012 37110-6009 37110-6023 37110-6005 37110-6018 37110-6004 37182-6001 37110-6021 37110-6003 37110-6011 37160-6001 37110-6002 37110-6006 37110-9002
58948-070CS 820.43 USD InStock
58948-070 58948-091 80089-636 58947-114 58947-136 58947-138 58948-230 58947-128 58947-106 58948-273 58948-251 58948-193 58948-171 58948-150 58948-218 58947-120 58947-132 58948-138 58947-122 58948-116 58948-025 58948-950 58947-124 58948-309
VWR® Spinbar® Magnetic Stir Bars, Octagon
Stirrers Magnetic Stirring Bars
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape.

  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low-friction coating; durable and inert
  • Pivot ring aids in reducing friction and chattering (58948-091 does not have a pivot ring)
  • Autoclavable
  • Manufactured in a registered ISO 9001:2008, as verified by SGS certification

Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars.

Ordering information: 6-Assortment includes six Octagon Spinbar® Magnetic Stir Bars, 2 each size; 5/16 x 1" (8 x 25.4mm), 5/16 x 1-1/2" (8 x 38.1mm); 5/16 x 2" (8mm x 50.8mm).
12-Assortment: Includes two of each 12.7x7.9mm (1/2x5/16"), 15.9x7.9mm (5/8x5/16"), 25.4x9.5mm (1x3/8"), 38.1x9.5mm (1-1/2x3/8"), 50.8x9.5mm (2x3/8"), and 63.5x9.5mm (2-1/2x3/8") sizes.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty