Would you like to log in again?
ELECTRODE HOLDER
Supplier: Bel-Art Products, a Part of SP
HOLDER, ELECTRODE^ The Electrode Holder snaps onto the end of the Flexible Arm affording 3 stepped wells for holding an electrode, a reference electrode, and an ATC probe.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ELECTRODE HOLDER 58948-296 F18317-1000 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty