Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Spinbar® Micro Stir Bars
Supplier: VWR
VWR® Spinbar®
37119-6011 37119-6183 37119-6003 37119-6001 37119-6010 37119-6008 37119-6005 37119-6004 37119-6007 37119-6009 37119-6002
76001-878CS 343.58 USD InStock
76001-878 76001-876 58948-976 76001-874 76001-872 58948-377 58948-411 58948-400 58948-375 58948-397 58948-353
VWR® Spinbar® Micro Stir Bars
Stirrers Magnetic Stirring Bars
Micro (Flea) magnetic stirring bars are designed for stirring small volumes in vessels such as vials, tubes, and gradient makers.
  • Particularly useful for environmental testing
  • Valuable for preparing and evaluating small sample volumes
  • Teflon® PTFE encapsulated
  • Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty