Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Pyrex® Spinbar® Stir Bars, Round, Glass
Supplier: VWR
VWR® Spinbar®
37101-6002 37101-6012 37101-6001 37101-6008
58948-488CS 2534.87 USD InStock
58948-488 58948-444 58948-466 58948-422
VWR® Pyrex® Spinbar® Stir Bars, Round, Glass
Stirrers Magnetic Stirring Bars
Pyrex® Spinbar® glass encapsulated Alnico magnet stirring bars are useful for high temperature applications up to 274°C (525°F).
  • Completely smooth surface offers “zero absorption”; easy to clean
  • Meets US Pharmacopoeia specifications for Type I Borosilicate glass
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty