Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Nalgene Suspended Magnetic Stir Bars, Thermo Scientific
Supplier: Thermo Scientific
DS6630-0250 DS6630-1000 DS6630-4000
58948-821CS 593.71 USD InStock
58948-821 58948-864 58948-782
Nalgene Suspended Magnetic Stir Bars, Thermo Scientific
Stirrers Magnetic Stirring Bars
Nalgene Floating Magnetic Stir Bars are designed for constant low-speed stirring action in tissue culture applications
The stir bars are ideal for tissue culture applications. The bar is slightly raised off the bottom surface of the vessel to minimize grinding effect on cells
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty