Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Spinbar® Magnetic Stir Bars, Polygon without Pivot Ring
Supplier: VWR
VWR® Spinbar®
37120-6058 37120-6012 37120-6003 37120-6002 37120-6078 37120-6005 37120-6001 37120-6038 37120-6004
58948-988CS 598.88 USD InStock
58948-988 58948-986 58948-080 58948-984 58948-985 58948-982 58948-983 58948-980 58948-981
VWR® Spinbar® Magnetic Stir Bars, Polygon without Pivot Ring
Stirrers Magnetic Stirring Bars
Polygon-shaped Alnico magnet encapsulated in FDA grade Teflon® PTFE is durable and inert.
  • Suitable for general stirring applications
  • Durable
  • Inert
Polygon-shaped stir bars come without a pivot ring.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty