Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Spinfin® Magnetic Stir Bars
Supplier: Avantor
VWR® Spinfin®
37125-6003 37125-6006 37125-6004 37125-6001 37125-6002 37125-6005
76001-868CS 567.83 USD InStock
76001-868 58948-996 58948-994 76001-870 58948-992 58948-990
VWR® Spinfin® Magnetic Stir Bars
Stirrers Magnetic Stirring Bars
Spinfin® magnetic stirring bars agitate liquids in round bottom vessels such as test tubes, cylinders and flasks, where other stirring bars cannot be used
  • Maintains its position in a vessel and will not wobble
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
Alnico magnet encapsulated in FDA grade Teflon® PTFE is durable and inert.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty