Would you like to log in again?
VWR® Spinfin® Double Stir Bars
Supplier: VWR
VWR® Double Spinfin®
37126-6002 37126-6003 37126-6001
58948-998CS 1005.99 USD InStock
58948-998 58949-004 58949-000
VWR® Spinfin® Double Stir Bars
Stirrers Magnetic Stirring Bars
Double Spinfin® Magnetic Stirring Bars agitate liquids in round bottom vessels such as test tubes, cylinders, and flasks, where other stirring bars cannot be used.
  • Fins on top and bottom ensure complete agitation
  • Maintains its position in a vessel and will not wobble
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low-friction coating; durable and inert
  • Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty