Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Spinstar® Magnetic Stir Bars
Supplier: VWR
VWR® Spinstar®
37146-6400 37146-6100 37146-6250
58949-028CS 2931.4 USD InStock
58949-028 58949-026 58949-032
VWR® Spinstar® Magnetic Stir Bars
Stirrers Magnetic Stirring Bars
Spinstar® magnetic stirring bars create a deep mixing vortex at relatively slow speeds
  • Star shape is designed to fit the inside diameter of most commonly-used beakers
  • Perfect for applications requiring slow, thorough mixing
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low friction coating; durable and inert.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty