Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Spinbar® Stir Bars, Octagon
Supplier: VWR
VWR® Spinbar®
37157-6020 37157-6010 37157-6005 37157-6015
58949-036CS 611.09 USD InStock
58949-036 58949-038 58949-034 58949-040
VWR® Spinbar® Stir Bars, Octagon
Stirrers Magnetic Stirring Bars
Convenient blister packs of PTFE resin-coated, Spinbar® magnetic stir bars.
  • PTFE resin coating
  • Alnico magnet
  • Most commonly used shape
Alnico magnets encapsulated in FDA grade Teflon® PTFE are durable and inert.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty