Would you like to log in again?
VWR® Spinwedge® Magnetic Stir Bars
Supplier: VWR
VWR® Spinwedge®
37123-6002 37123-6001 37123-6003 37123-6004
76001-896CS 406.64 USD InStock
76001-896 58949-254 58949-232 58949-265
VWR® Spinwedge® Magnetic Stir Bars
Stirrers Magnetic Stirring Bars
Spinwedge® magnetic stirring bars provide strong turbulence at fairly low speeds and are well suited for churning sediment or dissolving salts
  • Wedge shape is designed to plow up sediment for thorough dispersion
  • Sharply angled broad face provides strong mixing action, perfect for viscous solutions
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty