Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Spinvane® Stir Bars for Test Tubes, Micro Vials, and Conical Bottom Centrifuge Tubes
Supplier: VWR
VWR® Spinvane®
37134-9001 37134-6001 37136-6001 37135-6001 37137-6001
55999-000PK 777.65 USD InStock
55999-000 58949-274 58949-276 58949-270 58949-272
VWR® Spinvane® Stir Bars for Test Tubes, Micro Vials, and Conical Bottom Centrifuge Tubes
Stirrers Magnetic Stirring Bars
Solid Teflon® vane has a cross-mounted, molded-in magnet that can be trimmed to fit if needed without affecting the magnet.
  • Half-round shape is designed for 16 mm O.D. test tubes
  • Half-round tapered shape is designed for conical-bottom centrifuge tubes
  • Triangular shape is designed for small conical-shaped tubes and micro vials
  • Autoclavable
Each size is manufactured for a specific size tube; can be trimmed to fit if needed without affecting the magnet.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty