Would you like to log in again?
SHAFT 30IN 316SS 3/8INDIAMETER
Supplier: LIGHTNIN
Shaft^ Blade shaft for use with lab mixers. Made of type 316 stainless steel. Diameter: 9.5mm. Length: 76.2cm.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SHAFT 30IN 316SS 3/8INDIAMETER 58958-012 143726 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty