Would you like to log in again?
Two-Speed Lab Blenders, Waring
Supplier: Waring
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7011S 7011G 7011G
74790-002EA 878.79 USD InStock
74790-002 58977-103 58977-089
Two-Speed Lab Blenders, Waring
Homogenizers Standard Laboratory Blenders
This versatile laboratory blender is applicable for most lab work, providing no-load speeds of 18,000rpm and 22,000rpm
Available with either a heat-resistant glass, or stainless steel 1L container. Both have a handle and two-piece vinyl and styrene lid. Motor housing is epoxy-coated. Rated Amps: 3.6; 120V, 50/60Hz; 0.40HP. UL listed. Also available in heavy-duty versions.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty