Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Stainless Steel Container for Waring® Blenders, 360 ml, Eberbach

Supplier: Eberbach

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

E8581.00 E8580.00
58983-026EA 1978.03 USD InStock
58983-026 58983-004
Stainless Steel Container for Waring® Blenders, 360 ml, Eberbach
Homogenizers Standard Laboratory Blenders
Special purpose container for rapidly disintegrating and mixing small volumes of microorganisms and chemicals

Fits Waring® power units. Constructed from heavy gauge, spun, stainless steel. Total volume: 360mL; working volume: 25 to 250mL. Overall height: 17cm (63/4").

Ordering information: Complete with semi-micro blending assembly with stainless steel blades and choice of friction-fit stainless steel cover or stainless steel screw cover with 15.9mm (5/8") sampling plug and rubber “O” ring seals. Without motor base.

Order Now
 

ORDER

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty