Would you like to log in again?
CONTAINER COOL BASE SS 3.78L
Supplier: WARING PRODUCTS DIVISION
Cool Base Container^ 3.78L. Stainless steel container with jacketed base, allowing for water cooling of bearings during extended operation periods. Does not include lid or cover gasket.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CONTAINER COOL BASE SS 3.78L 58985-123 2610C Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty