Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Reversible Air-Powered Stirrer, Arrow
Supplier: Arrow Fastener
G-RV
59025-122EA 1687.9 USD InStock
59025-122
Reversible Air-Powered Stirrer, Arrow
Stirrers Overhead Stirrers
Explosion-proof reversible stirrer can be set to rotate in the direction you choose by simply connecting the air supply line to the right port for counter-clockwise rotation or the left port for clockwise rotation
Heavy-duty high-torque stirrers are recommended for batches up to 200L depending on viscosity. Constant speed (50–1000 rpm) is attained through a 7 to 1 heavy-duty gear reduction, minimizing speed fluctuations from changes in air pressure.
Unit includes a needle valve to allow variable mixing speed and a special muffler that ensures quiet operation.
Ordering information: Complete with detachable, corrosion-resistant, 30.5Lx1dia. cm (12x3/8") stainless steel shaft and 6.4dia. cm (21/2") propeller; 1.2m (4') air hose with double fittings; and 25.4Lx1.4dia. cm (10x9/16") mounting rod.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty