Would you like to log in again?
CHEMWARE® PTFE Fluoropolymer Stirring Rods, Saint Gobain Performance Plastics
Supplier: Saint-Gobain
Chemware®
D1069088 D1069089 D1069087 D1069091
59062-304EA 172.21 USD InStock
59062-304 59062-188 59062-213 59062-246
CHEMWARE® PTFE Fluoropolymer Stirring Rods, Saint Gobain Performance Plastics
Stirrers Stirring Rods
These stirring rods are designed with enclosed steel insert for stiffness but can be able bent to various shapes for use as a powered stirrer.
  • Heat resistant to 550 °F (288 °C)
  • Inert to highly corrosive reagents
  • Antistick properties helps to facilitate cleaning
  • Bendable into shape
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty