Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
SP Bel-Art Stir & Add™ Cuvette Mixers, Bel-Art Products, a part of SP
Supplier: Bel-Art
F37752-0000
59063-125CS 1026.29 USD InStock
59063-125
SP Bel-Art Stir & Add™ Cuvette Mixers, Bel-Art Products, a part of SP
Stirrers Stirring Rods
Made of clear acrylic with a straight handle, this tool is used when reactions are performed directly into cuvettes, to mix added reagents.
  • Clear acrylic construction
  • Straight handle
  • Complete mixing with simple up and down motion
Up and down motion in cell provides complete mixing. Shaft diameter is 3.17 mm (0.125").
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty