Would you like to log in again?
SP Bel-Art Stopcocks, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SCIENCEWARE®
F42040-0000
59097-102EA 75.04 USD InStock
59097-102
SP Bel-Art Stopcocks, Bel-Art Products, a part of SP
Stopcocks
Polypropylene plastic body has Teflon® PTFE plug, washer and bright red handle.
Taper fits on desiccator port, and the plug is 3 way for vacuum draw, shut-off and vacuum release.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty