Would you like to log in again?
High Vacuum Grease, DuPont
Supplier: DuPont
1658832 1597418 1658832 1597418
59344-066CS 5191.19 USD InStock
59344-066 59344-055 CA59344-055 CA59344-066
High Vacuum Grease, DuPont
Lubricants
High vacuum grease is ideal for sealing and lubricating vacuum and pressure systems
This silicone product has a low volatility to hold deep vacuums; it resists oxidation, is nonmelting and nongumming, and has good thermal stability. Grease is translucent white with a heavy consistency.
It can also be used to maintain laboratory equipment.
Certifications: Meets FDA CFR 21 175.300, USDA, NSF 61.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty