Would you like to log in again?
DECKMNT TURRET/4 NEED VLV/SHNK
Supplier: WATERSAVER FAUCET CO
Needle Valve^ Deck mounted 4-way turret base. Removable serrated hose end. Forged brass 4-arm handle and valve body. Removable/replaceable stainless steel seat and floating cone. Includes 9.5mm IPS mounting shank, locknut, and washer. Maximum working pressure: 125psi.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
DECKMNT TURRET/4 NEED VLV/SHNK 59382-020 L2880-134WS Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty