Would you like to log in again?
Needle Valves, WaterSaver Faucet
Supplier: WaterSaver Faucet
L2880-131WS L2880-146WS L2880-132AWS L2880-158WS L2880-132SWS
59382-016EA 266.55 USD InStock
59382-016 59382-018 59382-022 59382-024 59382-014
Needle Valves, WaterSaver Faucet
Faucets and Sinks Faucets
Straight pattern laboratory needle valve is for gas/air/vacuum service.
Complete with 9.5 mm (3/8") IPS mounting shank, locknut, and washer. Maximum working pressure of 125 psi. Please specify service when ordering.
It has a removable serrated hose end and forged brass four-arm handle and valve body. Stainless steel seat and floating cone are removable/replaceable for infinite service life.
Caution: Not for use with water.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty