Would you like to log in again?
PYREX® Polyethylene [ST] Stoppers, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PYREX®
7624-19 7624-27 7624-22 7624-32 7624-8 7624-13 7624-38 7624-16 7624-9
59492-121CS 44.01 USD InStock
59492-121 59492-187 59492-100 59492-123 59492-063 59492-085 59492-041 59492-125 59492-039
PYREX® Polyethylene [ST] Stoppers, Corning
Stoppers PE/LDPE/HDPE Stoppers
These PYREX® No.13 stoppers are made from linear HDPE and have closed bottoms to conform to Standard Taper stopper dimensions.
  • Blue and white with a closed bottom
  • Three rings on the plug provide an efficient seal
  • Improved design fully protects the neck from accidental tipping
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty