Would you like to log in again?
VWR® Micro Stoppers
Supplier: VWR
VWR®
M21
59580-001PK 824.26 USD InStock
59580-001
VWR® Micro Stoppers
Stoppers Rubber Stoppers
Select from soft red natural rubber or green neoprene for higher heat resistance.
  • Natural rubber or neoprene fabrication
The diameter tapers from 12.7mm (1/2") to 3mm (3/32"). Length: 50mm (2"). Length may be cut to fit openings from 4 to 12mm (5/32 to 15/32") in diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty