Would you like to log in again?
VWR STOPPER RUBBER 8.5 PK1
Supplier: VWR International
VWR* Black Rubber Stoppers, Solid^ High percentage of pure rubber; weigh less than neoprene; low in free sulfur (1/2%). Size: 8.5. Diameter of top: 43mm. Diameter of bottom: 36mm. Length: 25mm.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR STOPPER RUBBER 8.5 PK1 59580-320 Case of 5 Retrieving
Pack of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty