Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® One-Hole Rubber Stoppers, Black
Supplier: VWR
VWR®
12-M291 00-M291 5--M291 2--M291 11-M291 13-M291 7--M291 135M291 K8.5M291 14-M291 000M291 105M291 6--M291 9--M291 1--M291 9.5M291 10-M291 8--M291 0--M291 5.5M291 6.5M291 4--M291 3--M291 7.5M291 115M291
59581-200CS 662.84 USD InStock
59581-200 59581-243 59581-287 59581-265 59581-221 59581-120 59581-560 59581-582 59581-185 59581-163 59581-141 59581-480 59581-527 59581-549 59581-505 59581-108 59581-469 59581-403 59581-425 59581-447 59581-323 59581-389 59581-367 59581-301 59581-345
VWR® One-Hole Rubber Stoppers, Black
Stoppers Rubber Stoppers
These black high-percentage rubber stoppers are low in free sulfur and have a single hole to facilitate laboratory procedures.
  • Remain resilient and pliable under most laboratory conditions
Stoppers sized as 000 through 0 have a 3mm diameter hole; the size 1 stopper has a 4mm hole. Stoppers sized as 2 and larger have a 5mm diameter hole.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty