Would you like to log in again?
VWR® Two-Hole Rubber Stoppers, Black
Supplier: VWR
VWR®
8--M292 6.5M292 6--M292 10-M292 11-M292 7--M292 3--M292 2--M292 135M292 5--M292 4--M292 14-M292 9--M292 15-M292 12-M292
59582-406CS 811.18 USD InStock
59582-406 59582-326 59582-520 59582-541 59582-563 59582-268 59582-246 59582-202 59582-166 59582-144 59582-188 59582-440 59582-484 59582-280 59582-360
VWR® Two-Hole Rubber Stoppers, Black
Stoppers Rubber Stoppers
These black, high percentage rubber stoppers are low in free sulfur and have two holes to facilitate laboratory procedures.
  • Stoppers remain resilient and pliable under most laboratory conditions
  • Low free sulfur
Stoppers sized as 00 have 3mm diameter holes; the size 1 stopper has 4mm holes. Stoppers sized as 2 and larger have 5mm diameter holes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty