Would you like to log in again?
VWR® Pure Gum Rubber Stoppers, Solid
Supplier: VWR
VWR®
5--M180 6--M180 2--M180 00-M180 9--M180 12-M180 3--M180 4--M180 8--M180 0--M180 7--M180
59583-260CS 519.63 USD InStock
59583-260 59583-282 59583-329 59583-307 59583-340 59583-409 59583-147 59583-169 59583-205 59583-227 59583-249
VWR® Pure Gum Rubber Stoppers, Solid
Stoppers Rubber Stoppers
These solid, pure gum, amber rubber stoppers contain less free sulfur (max
  • Autoclavable
  • Soft and pliable under normal laboratory use
  • Excellent resilience
Retain their softness and pliability under normal laboratory use. Exhibit excellent resilience; withstand repeated autoclaving. Stoppers are available in whole sizes 00 through 12. Length: 25mm.
0.25%) and are less dense than standard black rubber stoppers with approximately 20% more stoppers per pound.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty