Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Pure Gum Rubber Stoppers, One-Hole
Supplier: VWR
VWR®
12-M181 4--M181 11-M181 7--M181 5--M181 8--M181 0--M181 2--M181 3--M181 10-M181 9--M181 6--M181
59584-219CS 698.63 USD InStock
59584-219 59584-194 59584-150 59584-172 59584-252 59584-274 59584-296 59584-230 59584-354 59584-332 59584-310 59584-117
VWR® Pure Gum Rubber Stoppers, One-Hole
Stoppers Rubber Stoppers
These highly resilient, pure gum, amber rubber stoppers have less free sulfur (max
  • High resiliency
  • Variety of stopper sizes
  • One-hole design
Added hole facilitates laboratory usage. Stoppers sized as 00 or 0 have a 3mm hole diameter; size 1 stoppers have a 4mm hole. Rubber stoppers sized as 2 and larger have a 5mm diameter hole.
0.25%) and lower density than standard black rubber stoppers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty