Would you like to log in again?
VWR® Pure Gum Rubber Stoppers, Two-Hole
Supplier: VWR
VWR®
4--M182 3--M182 0--M182 10-M182 9--M182 6--M182 12-M182 5--M182
59585-302CS 573.31 USD InStock
59585-302 59585-346 59585-288 59585-222 59585-200 59585-186 59585-164 59585-109
VWR® Pure Gum Rubber Stoppers, Two-Hole
Stoppers Rubber Stoppers
These highly resilient, pure gum, off-white rubber stoppers have less free sulfur (max
  • High resiliency
  • Variety of stopper sizes
  • Two-hole design
Made with two holes to facilitate laboratory usage. Stoppers sized as 00 or 0 have 3mm diameter holes; size 1 stoppers have 4mm holes. Rubber stoppers sized as 2 and larger have 5mm diameter holes. Length: 25mm.
0.25%) and are lower density than standard black rubber stoppers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty