Would you like to log in again?
VWR® Rubber Stoppers
Supplier: VWR
VWR®
TW-ASSORT TW-9 TW-7 TW-10 TW-3 TW-5 TW-8 TW-4 TW-6
59586-157CS 663.74 USD InStock
59586-157 59586-167 59586-159 59586-158 59586-164 59586-163 59586-160 59586-162 59586-161
VWR® Rubber Stoppers
Stoppers Rubber Stoppers
Versatile rubber stoppers are four stoppers in one.
  • Easy conversion between stopper types
  • Versatile design
  • Soft and resilient
  • Natural rubber construction
  • Autoclavable
Three simple twist-off seals permit easy conversion from a solid to a one, two, or three-hole stopper for all your laboratory needs. Soft, resilient, natural, white rubber demonstrates exceptional aging characteristics and withstands repeated autoclaving.
Ordering information: Stoppers are available in sizes 3 to 10 or as an assortment of 33 stoppers. Sizes 3 and 4 only have two twist-off seals. Stopper assortment 59586-167 contains nine size 3 stoppers, eight size 4, six size 5, four size 6, two size 7, two size 8, one size 9, and one size 10 stopper. For black stoppers (shown above), contact your Avantor representative.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty