Would you like to log in again?
Synthware Adapters, Vacuum or Argon, Kemtech America
Supplier: Kemtech America
Synthware
A291922 A291420 A292942 A292440
60000-162EA 26.35 USD InStock
60000-162 60000-168 60000-164 60000-166
Synthware Adapters, Vacuum or Argon, Kemtech America
Adapters Adapters, Joints with Tube Connectors
Use this adapter for vacuum or argon applications.
  • Inner [ST] joint
  • Side hose connection
Featuring a hose connection on the side and inner [ST] joint. Hose ext. Ø is 8 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty