Would you like to log in again?
Synthware Vacuum/Inert Gas Adapters, 90°, Kemtech America
Supplier: Kemtech America
A921922 A922440 A921420
60000-182EA 24.34 USD InStock
60000-182 60000-184 60000-180
Synthware Vacuum/Inert Gas Adapters, 90°, Kemtech America
Adapters Adapters, Joints with Tube Connectors
Adapters for vacuum/inert gas applications.
  • 90° angle
  • Top hose connection
  • Bottom male [ST] joint
Vacuum/inert gas adapters feature a 90° angle hose connection on the top and a male joint.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty