Would you like to log in again?
Synthware Vacuum Adapter, 90° with Outer Joint, Kemtech America
Supplier: Kemtech America
Synthware
A951420 A951922
60000-190EA 24.34 USD InStock
60000-190 60000-188
Synthware Vacuum Adapter, 90° with Outer Joint, Kemtech America
Adapters Adapters, Joints with Tube Connectors
Vacuum adapter features a 90° angle hose connection.
  • 90° angle
  • Top hose connection
  • Outer [ST] joint
Featuring a 90° angle hose connection on the top and an outer joint.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty