Would you like to log in again?
Synthware Vacuum Adapters with Glass Stopcock, Kemtech America
Supplier: Kemtech America
Synthware
A611922 A612440 A611420
60000-256EA 76.89 USD InStock
60000-256 60000-252 60000-254
Synthware Vacuum Adapters with Glass Stopcock, Kemtech America
Adapters Adapters, Joints with Tube Connectors
Use this vacuum adapter with glass stopcock for regulating the flow of gases or liquids into reaction systems.
  • [ST] joint
  • Glass stopcock
  • With hose connection
Featuring a stopcock bore of 2 mm. Hose connection size of 8 mm O.D. at the middle serration.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty